Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FLRT1 Antibody (017), Alexa Fluor™ 700 , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289894AF700
Description
FLRT1 Monoclonal antibody specifically detects FLRT1 in Human samples. It is validated for ELISASpecifications
FLRT1 | |
Monoclonal | |
Alexa Fluor 700 | |
fibronectin leucine rich transmembrane protein 1, Fibronectin-like domain-containing leucine-rich transmembrane protein 1, leucine-rich repeat transmembrane protein FLRT1, MGC21624 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human FLRT1 (Q9NZU1-1; Met1-Pro524). | |
0.1 mL | |
Cellular Markers, Extracellular Matrix, Mesenchymal Stem Cell Markers, Stem Cell Markers | |
23769 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
017 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction