Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FLRT1 Monoclonal Antibody (10B1A3)
GREENER_CHOICE

Catalog No. PIMA530438
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA530438 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA530438 Supplier Invitrogen™ Supplier No. MA530438
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human FLRT1.

This gene encodes a member of the fibronectin leucine rich transmembrane protein (FLRT) family. The family members may function in cell adhesion and/or receptor signalling. Their protein structures resemble small leucine-rich proteoglycans found in the extracellular matrix. The encoded protein shares sequence similarity with two other family members, FLRT2 and FLRT3. This gene is expressed in kidney and brain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FLRT1
Applications ELISA
Classification Monoclonal
Clone 10B1A3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene FLRT1
Gene Accession No. Q9NZU1
Gene Alias AW742165; D630040I23Rik; Fibronectin leucine rich transmembrane protein 1; fibronectin leucine rich transmembrane protein 3; fibronectin-like domain-containing leucine-rich transmembrane protein 1; Flrt1; Leucine-rich repeat transmembrane protein FLRT1; RGD1565152; SPG68; UNQ752/PRO1483
Gene Symbols FLRT1
Host Species Mouse
Immunogen Recombinant Human FLRT1 protein (Met1-Pro524).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23769
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.