Learn More
FLRT1 Monoclonal Antibody (4E10)

Description
FLRT1 Monoclonal Antibody for ELISA
Specifications
Specifications
| Antigen | FLRT1 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 4E10 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | FLRT1 |
| Gene Accession No. | Q9NZU1 |
| Gene Alias | AW742165; D630040I23Rik; Fibronectin leucine rich transmembrane protein 1; fibronectin leucine rich transmembrane protein 3; fibronectin-like domain-containing leucine-rich transmembrane protein 1; Flrt1; Leucine-rich repeat transmembrane protein FLRT1; RGD1565152; SPG68; UNQ752/PRO1483 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.