Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FLRT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584211
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584211 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584211 Supplier Invitrogen™ Supplier No. PA584211
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAIKDITSEM DECFETGPQG GVANAAAKTT ASNHASATTP QGSLFTLKAK Highest antigen sequence indentity to the following orthologs: Rat - 92%, Mouse - 92%.

This gene encodes a member of the fibronectin leucine rich transmembrane protein (FLRT) family. The family members may function in cell adhesion and/or receptor signalling. Their protein structures resemble small leucine-rich proteoglycans found in the extracellular matrix. The encoded protein shares sequence similarity with two other family members, FLRT2 and FLRT3. This gene is expressed in kidney and brain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FLRT1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FLRT1
Gene Accession No. Q9NZU1
Gene Alias AW742165; D630040I23Rik; Fibronectin leucine rich transmembrane protein 1; fibronectin leucine rich transmembrane protein 3; fibronectin-like domain-containing leucine-rich transmembrane protein 1; Flrt1; Leucine-rich repeat transmembrane protein FLRT1; RGD1565152; SPG68; UNQ752/PRO1483
Gene Symbols FLRT1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FLRT1. Recombinant protein control fragment (Product #RP-101256).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23769
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.