Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FLRT2 Antibody (11B5G9), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP306631R
Description
FLRT2 Monoclonal antibody specifically detects FLRT2 in Human samples. It is validated for ELISASpecifications
FLRT2 | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
11B5G9 | |
ELISA | |
Fibronectin Leucine Rich Transmembrane Protein 2, Fibronectin-Like Domain-Containing Leucine-Rich Transmembrane Protein 2, KIAA0405, Leucine-Rich Repeat Transmembrane Protein FLRT2 | |
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human FLRT2 (O43155; Met 1-Ser 539). | |
0.1 mL | |
Cell Biology, Signal Transduction, Stem Cells | |
23768 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction