Learn More
Invitrogen™ FLRT2 Monoclonal Antibody (367205)

Description
In direct ELISAs and Western blots, this antibody does not cross-react with recombinant human (rh) FLRT1 or rhFLRT3. Reconstitute at 0.5 mg/mL in sterile PBS.
Specifications
Specifications
| Antigen | FLRT2 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 367205 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | FLRT2 |
| Gene Accession No. | O43155 |
| Gene Alias | Fibronectin leucine rich transmembrane protein 2; fibronectin-like domain-containing leucine-rich transmembrane protein 2; FLRT2; Kiaa0405; Leucine-rich repeat transmembrane protein FLRT2; UNQ232/PRO265 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.