Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FLRT2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA532122
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA532122 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA532122 Supplier Invitrogen™ Supplier No. PA532122
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: U87-MG. Predicted reactivity: Mouse (100%), Rat (99%), Rhesus Monkey (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a member of the fibronectin leucine rich transmembrane protein (FLRT) family. FLRT family members may function in cell adhesion and/or receptor signalling. Their protein structures resemble small leucine-rich proteoglycans found in the extracellular matrix.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FLRT2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.08 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene FLRT2
Gene Accession No. O43155
Gene Alias Fibronectin leucine rich transmembrane protein 2; fibronectin-like domain-containing leucine-rich transmembrane protein 2; FLRT2; Kiaa0405; Leucine-rich repeat transmembrane protein FLRT2; UNQ232/PRO265
Gene Symbols FLRT2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the Extracellular domain of human FLRT2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23768
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.