Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FLRT2 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | FLRT2 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
FLRT2 Polyclonal specifically detects FLRT2 in Mouse samples. It is validated for Western Blot.Specifications
FLRT2 | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
23768 | |
IgG | |
Affinity purified |
Western Blot | |
Unconjugated | |
Rabbit | |
Mouse | |
Fibronectin Leucine Rich Transmembrane Protein 2, Fibronectin-Like Domain-Containing Leucine-Rich Transmembrane Protein 2, KIAA0405, Leucine-Rich Repeat Transmembrane Protein FLRT2 | |
A synthetic peptide of Human FLRT2. | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title