Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FLRT3 Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299728PCP
Description
FLRT3 Polyclonal antibody specifically detects FLRT3 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)Specifications
FLRT3 | |
Polyclonal | |
ELISA, Immunohistochemistry-Paraffin | |
fibronectin leucine rich transmembrane protein 3, Fibronectin-like domain-containing leucine-rich transmembrane protein 3, HH21, KIAA1469, leucine-rich repeat transmembrane protein FLRT3 | |
Produced in rabbits immunized with purified, recombinant Human FLRT3 (Accession#: NP_938205.1; Met 1-Pro 528) | |
0.1 mL | |
Signal Transduction | |
23767 | |
Store at 4C in the dark. | |
IgG |
ELISA, Immunohistochemistry (Paraffin) | |
PerCP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction