Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Flt-3/Flk-2/CD135 Antibody (12B6), Biotin, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NB11094155B
Description
Flt-3/Flk-2/CD135 Monoclonal specifically detects Flt-3/Flk-2/CD135 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Flt-3/Flk-2/CD135 | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
FLT3 | |
Mouse Flt-3/Flk-2/CD135 (extracellular domain) | |
0.1 mL | |
Primary | |
This recognizes mouse Flt3/CD135 Antibody (12B6) in western blot. | |
Store at 4C in the dark. | |
IgG2 |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
12B6 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
CD135, CD135 antigen, EC 2.7.10, fetal liver kinase 2, FL cytokine receptor, FLK2, FLT-3, FLT3 receptor tyrosine kinase, Fms-like tyrosine kinase 3, fms-related tyrosine kinase 3, growth factor receptor tyrosine kinase type III, Stem cell tyrosine kinase 1, STK-1, STK1EC 2.7.10.1, Tyrosine-protein kinase receptor FLT3 | |
Rat | |
Protein G purified | |
Cell Cycle and Replication, Phospho Specific | |
2322 | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction