Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Flt-3/Flk-2/CD135 Antibody (A2F10), Alexa Fluor™ 594, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP143352AF594
Description
Flt-3/Flk-2/CD135 Monoclonal specifically detects Flt-3/Flk-2/CD135 in Human, Mouse samples. It is validated for Flow Cytometry.Specifications
Flt-3/Flk-2/CD135 | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
FLT3 | |
The immunogen for this antibody was CD135. | |
0.1 mL | |
Primary | |
Human, Mouse | |
IgG2a κ |
Flow Cytometry | |
A2F10 | |
Flow Cytometry | |
CD135, CD135 antigen, EC 2.7.10, fetal liver kinase 2, FL cytokine receptor, FLK2, FLT-3, FLT3 receptor tyrosine kinase, Fms-like tyrosine kinase 3, fms-related tyrosine kinase 3, growth factor receptor tyrosine kinase type III, Stem cell tyrosine kinase 1, STK-1, STK1EC 2.7.10.1, Tyrosine-protein kinase receptor FLT3 | |
Rat | |
Protein A or G purified | |
Cell Cycle and Replication, Neuroscience, Neurotransmission, Phospho Specific, Protein Kinase | |
2322.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction