Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Flt-3/Flk-2/CD135 Antibody (MM0294-9G23) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$206.50
Specifications
Antigen | Flt-3/Flk-2/CD135 |
---|---|
Clone | MM0294-9G23 |
Concentration | LYOPH |
Applications | CyTOF, Western Blot, Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB019164
![]() |
Novus Biologicals
NBP2123110.025MG |
0.025 mg |
Each for $206.50
|
|
|||||
NBP212311
![]() |
Novus Biologicals
NBP212311 |
0.1 mg | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
Flt-3/Flk-2/CD135 Monoclonal specifically detects Flt-3/Flk-2/CD135 in Human samples. It is validated for Western Blot.Specifications
Flt-3/Flk-2/CD135 | |
LYOPH | |
Monoclonal | |
Purified | |
RUO | |
Cynomolgus Monkey, Human | |
CD135, CD135 antigen, EC 2.7.10, fetal liver kinase 2, FL cytokine receptor, FLK2, FLT-3, FLT3 receptor tyrosine kinase, Fms-like tyrosine kinase 3, fms-related tyrosine kinase 3, growth factor receptor tyrosine kinase type III, Stem cell tyrosine kinase 1, STK-1, STK1EC 2.7.10.1, Tyrosine-protein kinase receptor FLT3 | |
FLT3 | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
MM0294-9G23 | |
CyTOF, Western Blot, Flow Cytometry | |
Unconjugated | |
Mouse | |
Cell Cycle and Replication | |
P36888 | |
2322 | |
IgG1 | |
Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title