Learn More
Promega FLT1 Kinase Enzyme System, 1mg
Catalog No. PRV3002 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV3002 1 mg
1 options
Catalog No. PRV3002 Supplier Promega Supplier No. V3002
Add to Cart
Add to Cart
Request a Quote
Recombinant human FLT1 (amino acids 784-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. FLT1, also known as fms-like tyrosine kinase 1, is a member of the VEGFR family and is related to the oncogene ROS.
  • Easily Screen and Profile FLT1 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C