Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega FLT3 (D835Y) Kinase Enzyme System, 10microg

Catalog No. PRV4514 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV4514 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRV4514 Supplier Promega Supplier No. V4514
Add to Cart
Add to Cart

Recombinant human FLT3 (D835Y) (571-993) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.

  • Easily Screen and Profile FLT3 (D835Y) Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity

Specifications

Quantity 10 μg
Content And Storage <-65°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.