Learn More
Promega FLT3 Kinase Enzyme System, 10microg
Catalog No. PRV4064 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
PRV4064 10 μg
1 options
Catalog No. PRV4064 Supplier Promega Supplier No. V4064
FLT3 is a receptor tyrosine kinase that has been shown to play a role in proliferation and survival of hematopoietic progenitor cells as well as differentiation of early B lymphoid progenitors.
  • Easily Screen and Profile FLT3 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 10 μg
Content And Storage <-65°C