Learn More
Promega FLT3 Kinase Enzyme System, 1mg
Catalog No. PRV6394 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV6394 1 mg
1 options
Catalog No. PRV6394 Supplier Promega Supplier No. V6394
Add to Cart
Add to Cart
Request a Quote
FLT3 is a receptor tyrosine kinase that has been shown to play a role in proliferation and survival of hematopoietic progenitor cells as well as differentiation of early B lymphoid progenitors.
  • Easily Screen and Profile FLT3 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C