Learn More
Invitrogen™ FLT3 Monoclonal Antibody (BV10A4), PE

Description
This antibody reacts with an extracellular epitope of CD135 (FLT3, FLK2, STK-1), a 130-160 kDa type I transmembrane receptor tyrosine kinase that is involved in early steps of hematopoiesis. This antibody will not cross-react with mouse.
Specifications
Specifications
| Antigen | FLT3 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | BV10A4 |
| Concentration | 0.04 mg/mL |
| Conjugate | PE |
| Formulation | PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 |
| Gene | FLT3 |
| Gene Accession No. | P36888 |
| Gene Alias | B230315G04; CD135; CD135 antigen; fetal liver kinase 2; Fetal liver kinase-2; FL cytokine receptor; FLK2; Flk-2; FLT3; Flt-3; fms related tyrosine kinase 3; FMS-like tyrosine kinase 3; fms-related tyrosine kinase 3; growth factor receptor tyrosine kinase type III; Ly72; OTTHUMP00000214829; Receptor-type tyrosine-protein kinase FLT3; RP11-153M24.3; stem cell tyrosine kinase 1; STK1; STK-1; tyrosine-protein kinase FLT3; tyrosine-protein kinase receptor flk-2; wmfl |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.