Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FLT3 Mouse anti-Human, Clone: OTI7D6, lyophilized, TrueMAB™
SDP

Catalog No. 501668659
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-166-8659 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-166-8659 Supplier OriGene Supplier No. CF808157
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO

FLT3 (FMS-like tyrosine kinase 3) is a member of the class III receptor tyrosine kinase family, and its internal tandem duplication (ITD) is preferentially seen in acute myeloid leukemia and myelodysplastic syndrome among various hematological maligcies. FLT3 is a membrane bound glycosylated protein that is expressed primarily in early hematopoietic progenitor cells, and play an important role in hematopoietic stem/progenitor cell survival and proliferation. Upon binding its ligand FL, FLT3 activates multiple signaling pathways including the PI3K/Akt and Ras/MEK/MAPK pathways. In cells expressing FLT3-ITD, these pathways are constitutively on, resulting in abnormal cell growth and anti-apoptotic activity. Mutations in the FLT3 protein are implicated in 1/3 of acute myeloid leukemia (AML) patients.

Specifications

Antigen FLT3
Applications Western Blot
Classification Monoclonal
Clone OTI7D6
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene FLT3
Gene Accession No. P36888
Gene Alias CD135, FLK-2, FLK2, STK1
Gene Symbols FLT3
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 169-444 of human FLT3 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2322
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.