Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FLT3 Mouse anti-Human, Clone: OTI7D6, liquid, TrueMAB™
SDP

Catalog No. 501681937
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-1937 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-1937 Supplier OriGene Supplier No. TA808157
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FLT3 (FMS-like tyrosine kinase 3) is a member of the class III receptor tyrosine kinase family, and its internal tandem duplication (ITD) is preferentially seen in acute myeloid leukemia and myelodysplastic syndrome among various hematological maligcies. FLT3 is a membrane bound glycosylated protein that is expressed primarily in early hematopoietic progenitor cells, and play an important role in hematopoietic stem/progenitor cell survival and proliferation. Upon binding its ligand FL, FLT3 activates multiple signaling pathways including the PI3K/Akt and Ras/MEK/MAPK pathways. In cells expressing FLT3-ITD, these pathways are constitutively on, resulting in abnormal cell growth and anti-apoptotic activity. Mutations in the FLT3 protein are implicated in 1/3 of acute myeloid leukemia (AML) patients.

Specifications

Antigen FLT3
Applications Western Blot
Classification Monoclonal
Clone OTI7D6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene FLT3
Gene Accession No. P36888
Gene Alias CD135, FLK-2, FLK2, STK1
Gene Symbols FLT3
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 169-444 of human FLT3 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2322
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.