Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FLT3LG Recombinant Rabbit Monoclonal Antibody (008), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
$293.50 - $565.00
Specifications
Antigen | FLT3LG |
---|---|
Clone | 008 |
Concentration | 1 mg/mL |
Applications | ELISA |
Classification | Recombinant Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA543449
|
Invitrogen™
MA543449 |
50 μL |
Each for $293.50
|
|
|||||
PIMA543098
|
Invitrogen™
MA543098 |
100 μL |
Each for $565.00
|
|
|||||
Description
Specificity: Human Flt3 Ligand/FLT3LG
FLT3LG (Fms Related Tyrosine Kinase 3 Ligand) is a Protein Coding gene. Diseases associated with FLT3LG include Aplastic Anemia and Brain Cancer. Among its related pathways are Hematopoietic Stem Cell Differentiation Pathways and Lineage-specific Markers and Ras signaling pathway. Gene Ontology (GO) annotations related to this gene include protein homodimerization activity and cytokine activity. Dendritic cells (DCs) provide the key link between innate and adaptive immunity by recognizing pathogens and priming pathogen-specific immune responses. FLT3LG controls the development of DCs and is particularly important for plasmacytoid DCs and CD8-positive classical DCs and their CD103-positive tissue counterparts.Specifications
FLT3LG | |
1 mg/mL | |
Recombinant Monoclonal | |
Liquid | |
RUO | |
PBS with no preservative | |
FL; Flt 3 ligand; Flt3; Flt3 L; Flt3 ligand; Flt3l; Flt3lg; fms related receptor tyrosine kinase 3 ligand; fms related tyrosine kinase 3 ligand; FMS-like tyrosine kinase 3 ligand; fms-related tyrosine kinase 3; fms-related tyrosine kinase 3 ligand; H-FLT-3-L; Ly72L; M-FLT-3-L; NEWGENE_2322792; SL cytokine | |
FLT3LG | |
IgG | |
Protein A | |
Antibody |
008 | |
ELISA | |
Unconjugated | |
Rabbit | |
Human | |
P49771 | |
2323 | |
Recombinant Human Flt3 Ligand/FLT3LG Protein. | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
FLT3LG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title