Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FLT4 Mouse anti-Human, Clone: 44CT92.4.4, Abnova™

Catalog No. 89934152 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-934-152 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-934-152 Supplier Abnova Corporation Supplier No. MAB12327
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant human FLT4.

This gene encodes a tyrosine kinase receptor for vascular endothelial growth factors C and D. The protein is thought to be involved in lymphangiogenesis and maintenance of the lymphatic endothelium. Mutations in this gene cause hereditary lymphedema type IA. [provided by RefSeq]

Specifications

Antigen FLT4
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone 44CT92.4.4
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant human FLT4.
Dilution Immunohistochemistry (1:10-1:50) Western Blot (1:2000) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene Alias FLT41/LMPH1A/PCL/VEGFR3
Gene Symbols FLT4
Host Species Mouse
Immunogen Recombinant His fusion protein corresponding to human FLT4.
Purification Method Protein G purification
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2324
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.