Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Flt4, Rat anti-Mouse, Clone: AFL4, Abnova™

Catalog No. 89999043 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-999-043 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-999-043 Supplier Abnova Corporation Supplier No. MAB16245
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Specifications

Antigen Flt4
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone AFL4
Conjugate Unconjugated
Dilution Flow Cytometry Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Immunohistochemistry (Frozen sections) Immunoprecipitation Western Blot The optimal working dilution should be determined by the end user.
Formulation In aqueous buffer (Stabilizer, 0.09% sodium azide).
Gene FMS-like tyrosine kinase 4
Gene Alias AI323512/Chy/Flt-4/VEGFR-3/VEGFR3
Gene Symbols Flt4
Host Species Rat
Immunogen Mouse Flt4.
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14257
Target Species Human, Mouse
Content And Storage Store at 4°C.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.