Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Fludarabine
GREENER_CHOICESDP

Catalog No. 349550 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-955-0 50 mg
34-951-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 34-955-0 Supplier R&D Systems™ Supplier No. 3495/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Purine analog; inhibits DNA synthesis

Purine analog that inhibits DNA synthesis. Exhibits antiproliferative activity (IC50 = 1.54 μM in RPMI cells) and triggers apoptosis through increasing Bax and decreasing Bid, XIAP and survivin expression. Inhibits cytokine-induced activation of STAT1 and STAT1-dependent gene transcription in lymphocytes. Also displays anticancer activity against hematological malignancies in vivo.

Chemical Identifiers

CAS 21679-14-1
Molecular Formula C10H12FN5O4

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 285.23
Purity >99%
Molecular Weight (g/mol) 285.24
Chemical Name or Material Fludarabine
Recommended Storage Desiccate at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.