Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Fluid Thioglycollate Medium with Indicator, Dextrose Medium

Catalog No. p-4523848
Click to view available options
Quantity:
10/Pk.
100/Pk.
20/Pk.
Format:
16mm x 125mm Tube
16mm x 125mm Tube, Hungate Cap
20mm x 113mm Tube
Borosilicate Bottle - Septum
Bottle
SB
Tube
WMB Lug
Media Type:
Fluid Thioglycollate Medium
Manufacturing Origin:
United States
This item is not returnable. View return policy
This item is not returnable. View return policy

Reduce prep time while isolating aerobes, anaerobes and microaerophilic organisms with this liquid medium with formats available that meet international industry guidelines.

Cultivate and isolate aerobes, anaerobes and microaerophilic organisms using Thermo Scientific™ Fluid Thioglycollate Medium with Indicator, Dextrose Medium.

Remove the need for pipetting, dispensing or autoclaving with convenient, pre-filled and ready-to-use tubes and bottles of Fluid Thioglycollate Medium w/Indicator, Dextrose.

Fluid Thioglycollate Medium is available as dehydrated culture medium or in ready-to-use prepared media tubes and bottles and in formats that meet international industrial guidelines for microbiological testing, including United States Pharmacopeia (USP) guidelines for sterility testing.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance AOAC, BAM, COMPF
Color Clear broth w/visible agar light to medium amber
Content And Storage 2 to 25°C
Format Tube
Manufacturing Origin United States
Media Type Fluid Thioglycollate Medium
Product Type Microbiology Media
Fill Volume 9 mL
Includes w/ Indicator, Dextrose Medium
Packaging Type Glass Tube
Quantity 100/Pk.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.