Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Flumequine, 98%

Click to view available options
Quantity:
1g
5g
25g
42835-25-6
C14H12FNO3
261.252
MFCD00079298
DPSPPJIUMHPXMA-UHFFFAOYSA-N
9-Fluoro-1,5,6,7-tetrahydro-5-methyl-1-oxopyrido[3,2,1-ij]quinoline-2-carboxylic Acid
3374
CHEBI:85269
CC1CCC2=C3N1C=C(C(=O)C3=CC(=C2)F)C(=O)O
42835-25-6
C14H12FNO3
261.252
MFCD00079298
DPSPPJIUMHPXMA-UHFFFAOYSA-N
9-Fluoro-1,5,6,7-tetrahydro-5-methyl-1-oxopyrido[3,2,1-ij]quinoline-2-carboxylic Acid
3374
CHEBI:85269
CC1CCC2=C3N1C=C(C(=O)C3=CC(=C2)F)C(=O)O

Inhibits topoisomerases in gram negative bacteria

Beilstein 490724
Merck Index 14,4137
Solubility Information Soluble in DMSO and dilute alkali hydroxides; Insoluble in water
Formula Weight 261.25
Physical Form Crystalline Powder
Percent Purity 98%
Color White
Melting Point 253°C to 255°C
Quantity 1g
Chemical Name or Material Flumequine

RTECSNumber : DK1672000