Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Flunixin Meglumine, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18602754
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
1 kg
10 kg
Packaging:
Amber Glass Bottle

F1293, 42461-84-7, C14H11F3N2O2·C7H17NO5

Spectrum™ Chemical Flunixin Meglumine, USP is a nonsteroidal, anti-inflammatory drug and antipyretic. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 42461-84-7
Molecular Formula C21H28F3N3O7
Molecular Weight (g/mol) 491.46
InChI Key MGCCHNLNRBULBU-WZTVWXICSA-N
IUPAC Name (2R,3R,4R,5S)-6-(methylamino)hexane-1,2,3,4,5-pentol; 2-{[2-methyl-3-(trifluoromethyl)phenyl]amino}pyridine-3-carboxylic acid
SMILES CNC[C@H](O)[C@@H](O)[C@H](O)[C@H](O)CO.CC1=C(NC2=NC=CC=C2C(O)=O)C=CC=C1C(F)(F)F

Specifications

CAS Max % 100%
Ignition Residue 0.2%
Loss on Drying 0.5%
Melting Range 135°C to 140°C
Packaging Amber Glass Bottle
Quantity 100 g
UN Number UN2811
Specific Rotation -9° to -12°
Percent Purity 98 to 102%
Grade USP
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Flunixin Meglumine, USP
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.