Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fluoboric Acid, Reagent, 48-50%, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Click to view available options
Quantity:
500 mL
4 L
6 x 500 mL
20 L
4 x 4 L
200 L
Packaging:
Drum
Poly Bottle
Poly Drum
Description
Spectrum™ Chemical Fluoboric Acid, Reagent , is an inorganic compound that is named oxonium tetrafluoroboranuide, is classified as a strong acid that is corrosive and is used in organic chemistry as a catalyst as well as in metal plating where it is combined with organic reagents and serves as an electrolyte in the cathode treatment of the plating process. Its reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.
Specifications
Specifications
| CAS Max % | 100% |
| Packaging | Poly Drum |
| Quantity | 20 L |
| UN Number | UN1775 |
| Percent Purity | 48 to 50% |
| Grade | Reagent |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction