Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FluoCells™ Prepared Slide #2 (BPAE cells with Mouse Anti-α-tubulin, AlexaFluor™ 488, FL Goat Anti-Mouse IgG, Texas Red™-X Phalloidin, and DAPI)

Catalog No. f14781
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F14781 1 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F14781 Supplier Invitrogen™ Supplier No. F14781
Add to Cart
Edge
Add to Cart

Multilabeled cell preparations for observation by fluorescence microscopy

FluoCells prepared slide #2 contains bovine pulmonary artery endothelial cells (BPAEC). Texas Red-X phalloidin is labeling the F-actin, anti—bovine α-tubulin mouse monoclonal 236-10501 in conjunction with BODIPY FL goat anti—mouse IgG antibody is labeling microtubules, and DAPI is labeling the nucleus.

Cell Analysis, Cellular Imaging, Immunofluorescence (IF)

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Type Prepared Microscope Slide
Product Line BODIPY FluoCells Texas Red
For Use With (Application) Immunofluorescence
Content And Storage Store at room temperature and protect from light.
Quantity 1 Each
Shipping Condition Room Temperature
Do you have any slides of pre-labeled cells or tissues that I can use to demonstrate with?

Some of the most popular are the FluoCells demonstration slides, which have cells or tissue sections that are labeled with three different dyes. FluoCells 1 and 2 have cultured cells, while FluoCells 3 is a tissue section (mouse kidney). Slides with fluorescent beads are also available.

DAPI and Hoechst dyes are quite similar to each other. Why would I choose one over the other?

DAPI is a very common blue-fluorescent dye for nuclear counterstaining and gives very bright labeling on nuclei in fixed and permeabilized cells and tissues. However, it is considered to be a semi-permeant to impermeant stain and provides inconsistent staining of live cells. Hoechst 33342 dye is cell-permeant and stains with the same binding mechanism and fluorescent color; it is preferred for live-cell imaging and is just as good as DAPI for fixed cell labeling.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.