Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluocinonide, Micronized, USP, 97-103%, Spectrum™ Chemical
SDP

Catalog No. 18602774
Click to view available options
Quantity:
250 mg
1 g
5 g
25 g
100 g
250 g
Packaging:
Amber Glass Bottle

F1123, 356-12-7, C26H32F2O7

Spectrum™ Chemical Fluocinonide, Micronized, USP is glucocorticoid steroid used as an anti-inflammatory treatment for disorders like seborrheic dermatitis or eczema. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 356-12-7
Molecular Formula C26H32F2O7
Molecular Weight (g/mol) 494.53
InChI Key WJOHZNCJWYWUJD-IUGZLZTKSA-N
IUPAC Name 2-[(1S,2S,4R,8S,9S,11S,12R,13S,19S)-12,19-difluoro-11-hydroxy-6,6,9,13-tetramethyl-16-oxo-5,7-dioxapentacyclo[10.8.0.02,9.04,8.013,18]icosa-14,17-dien-8-yl]-2-oxoethyl acetate
SMILES CC(=O)OCC(=O)[C@@]12OC(C)(C)O[C@@H]1C[C@H]1[C@@H]3C[C@H](F)C4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]21C

Specifications

CAS Max % 100%
Melting Point 308 to 311°C
Loss on Drying 1%
Packaging Amber Glass Bottle
Quantity 250 g
UN Number UN2811
Specific Rotation +81° to +89°
Percent Purity 97 to 103%
Grade USP
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Fluocinonide
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.