Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluorescamine, MP Biomedicals™

Catalog No. ICN19518383 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
1 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19518383 250 mg
ICN19518325 25 mg
ICN19518380 100 mg
ICN19518391 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN19518383 Supplier MP Biomedicals, Inc Supplier No. 0219518383
Add to Cart
Add to Cart

Chemical Identifiers

CAS 38183-12-9
Molecular Formula C17H10O4
Molecular Weight (g/mol) 278.26
MDL Number MFCD00005928
InChI Key ZFKJVJIDPQDDFY-UHFFFAOYNA-N
Synonym 4-phenylspiro[furan-2(3H)-1'-(3'-H)-isobenzofuran]-3, 3'-dione
PubChem CID 37927
SMILES O=C1OC2(OC=C(C2=O)C2=CC=CC=C2)C2=CC=CC=C12

Specifications

IUPAC Name 4'-phenyl-3H,3'H-spiro[2-benzofuran-1,2'-furan]-3,3'-dione
Formula Weight 278.3
Color White
Quantity 250 mg
Physical Form Powder
Chemical Name or Material Fluorescamine, For TLC analysis
EINECSNumber 253-814-5
Recommended Storage Store at Room Temperature (15 to 30°C).
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.