Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fluorescein Antibody (4-4-20 (enhanced)), Alexa Fluor™ 488, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP281026AF488
Description
Fluorescein Monoclonal specifically detects Fluorescein in Non-species specific samples. It is validated for Western Blot, Flow Cytometry.Specifications
| Fluorescein | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A purified | |
| RUO | |
| Fluorescein labelled proteins. | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, Flow Cytometry | |
| 4-4-20 (enhanced) | |
| Western Blot, Flow Cytometry | |
| FITC, fluorescein isothyocianate | |
| Fluorescein I-bovine serum albumin (FlI-BSA). | |
| 0.1 mL | |
| Primary | |
| Non-species specific | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction