Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fluorescein Antibody (4-4-20 (enhanced)) - Chimeric - Azide and BSA Free, Novus Biologicals™

Rabbit Monoclonal Antibody
$269.00 - $584.50
Specifications
Antigen | Fluorescein |
---|---|
Clone | 4-4-20 (enhanced) |
Dilution | Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, CyTOF |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP25263825
![]() |
Novus Biologicals
NBP2526380.025MG |
0.025 mg |
Each for $269.00
|
|
|||||
NBP25263802
![]() |
Novus Biologicals
NBP2526380.2MG |
0.2 mg |
Each for $584.50
|
|
|||||
Description
Fluorescein Monoclonal specifically detects Fluorescein in Non-species specific samples. It is validated for Western Blot, Flow Cytometry, Control, CyTOF-ready.Specifications
Fluorescein | |
Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
PBS with 0.02% Proclin 300. with 0.02% Proclin 300 | |
Fluorescein I-bovine serum albumin (FlI-BSA). | |
Primary | |
Store at 4C for up to 3 months. For longer storage, aliquot and store at -20C. |
4-4-20 (enhanced) | |
Western Blot, Flow Cytometry, CyTOF | |
Unconjugated | |
Rabbit | |
Non-species specific | |
FITC, fluorescein isothyocianate | |
IgG | |
Protein A or G purified | |
Fluorescein labelled proteins. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title