Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fluorescein Antibody (4-4-20 (enhanced)), DyLight 550, Novus Biologicals™

Armenian Hamster Monoclonal Antibody
Supplier: Novus Biologicals NBP281025R
Description
Fluorescein Monoclonal specifically detects Fluorescein in Non-species specific samples. It is validated for Western Blot, Flow Cytometry.Specifications
Fluorescein | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate | |
Armenian Hamster | |
Protein A purified | |
RUO | |
Fluorescein labelled proteins. | |
Store at 4°C in the dark. | |
IgG κ |
Western Blot, Flow Cytometry | |
4-4-20 (enhanced) | |
Western Blot, Flow Cytometry | |
FITC, fluorescein isothyocianate | |
Fluorescein I-bovine serum albumin (FlI-BSA). | |
0.1 mL | |
Primary | |
Non-species specific | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction