Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluorescein Antibody (4-4-20 (enhanced)), Janelia Fluor™ 646 - Chimeric, Novus Biologicals™
SDP

Rabbit Monoclonal Antibody

Supplier:  Novus Biologicals NBP252638JF646

Catalog No. NBP52638646


Only null left
Add to Cart

Description

Description

Fluorescein Monoclonal specifically detects Fluorescein in Non-species specific samples. It is validated for Western Blot, Flow Cytometry.
Specifications

Specifications

Fluorescein
Monoclonal
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
Rabbit
Protein A or G purified
Primary
Non-species specific
Purified
Flow Cytometry
4-4-20 (enhanced)
Flow Cytometry
FITC, fluorescein isothyocianate
Fluorescein I-bovine serum albumin (FlI-BSA).
0.1 mL
Fluorescein labelled proteins.
Store at 4°C in the dark.
IgG κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.