Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fluorescein Antibody, HRP, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB1206656
Description
Fluorescein Polyclonal specifically detects Fluorescein in Non-species specific samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Fluorescein | |
Polyclonal | |
Western Blot 1:5000-1:10000, ELISA 1:200000, Immunohistochemistry 1:500-1:2500, Immunohistochemistry-Paraffin 1:500-1:2500, Dot Blot | |
FITC, fluorescein isothyocianate | |
Fluorescein conjugated to Goat IgG | |
0.2 mg | |
Primary | |
Non-species specific | |
Purified |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Dot Blot | |
HRP | |
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free with 0.01% Gentamicin Sulfate | |
Goat | |
Immunogen affinity purified | |
RUO | |
Fluorescein | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction