Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Fluorescein, Disodium Salt
SCI_ED

Catalog No. p-4416716 Shop All Fisherbrand Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Powder
This item is not returnable. View return policy
This item is not returnable. View return policy
Stain and Indicator

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 518-47-8
Molecular Formula C20H10Na2O5
Molecular Weight (g/mol) 376.28
MDL Number MFCD00167039
InChI Key NJDNXYGOVLYJHP-UHFFFAOYSA-L
Synonym Uranin, Disodium Salt
IUPAC Name disodium 2-(6-oxido-3-oxo-3H-xanthen-9-yl)benzoate
SMILES [Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=CC(=O)C=C2OC2=CC([O-])=CC=C12

Specifications

Assay Percent Range 100% w/v
Linear Formula C20H10O5Na2
Formula Weight 412.3
Packaging Powder
Color Orange/Rust
Quantity 25 g
Chemical Name or Material Fluorescein, Disodium Salt
ShelfLife 5 years
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.