Learn More
Tocris Bioscience™ Fluorescein-NAD+
Description
This product is a replacement for R&D Systems product 4673-500-01
81 μg is supplied as 250 μl of a 0.25 mM solution in water.
Specifications
Specifications
| Chemical Name or Material | β-Nicotinamide-N6-[2-[[6-[fluorescein]-amino]-2-oxohexyl-hexyl]amino]-2-oxoethyl]adenine dinucleotide |
| Quantity | 81 μg |
| Target | Enzyme Probes and Enzyme Substrates |
| Molecular Formula | C56H64N10O22P2 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.