Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluorescein Sodium Salt, indicator, 98.5 to 100.5%, Solstice

Catalog No. 60045781 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
1 kg
4 x 1 kg
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-045-781 100 g Glass Bottle
60-045-782 1 kg Glass Bottle
60-045-783 4 x 1 kg Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60-045-781 Supplier Solstice Supplier No. 28803100G
Add to Cart
Add to Cart

Indicator, 98.5-100.5%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 518-47-8
Molecular Formula C20H10Na2O5
Molecular Weight (g/mol) 376.28
MDL Number MFCD00167039
InChI Key NJDNXYGOVLYJHP-UHFFFAOYSA-L
IUPAC Name disodium 2-(6-oxido-3-oxo-3H-xanthen-9-yl)benzoate
SMILES [Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=CC(=O)C=C2OC2=CC([O-])=CC=C12

Specifications

Melting Point >300°C
UN Number NONH for all modes of transport
Beilstein 3833041
Formula Weight 376.27g/mol
Percent Purity 98.5 to 100.5%
Grade Indicator
Packaging Glass Bottle
Color Red
Quantity 100 g
Physical Form Powder
Chemical Name or Material Fluorescein Sodium Salt
Show More Show Less
EINECSNumber 208-253-0
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.