Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluorescein Sodium Salt, Sigma-Aldrich

Catalog No. 01258812 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-812 100 g
01-258-969 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-258-812 Supplier MilliporeSigma Supelco Supplier No. F6377100G
Add to Cart
Add to Cart

Water soluble fluorescent dye used in permeability and vascular perfusion studies due to its small molecular radius. Sodium fluorescein is used as a fluorescent tracer to study blood-brain barrier (BBB) and blood-spinal cord barrier (BSCB) permeability in rodent models. The dye was used as a probe substrate in the study of organic anion transporting polypeptide (OATP) mediated drug transport in hepatocytes. Sodium fluorescein was also used to monitor in situ solute transportation into bone through fluorescence recovery after photobleaching (FRAP).

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 518-47-8
Molecular Weight (g/mol) 376.27 g/mol

Specifications

Color Orange to Red
pH 8.3
Quantity 100 g
Chemical Name or Material Fluorescein sodium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.