Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluorescein Sodium, Spectrum™ Chemical
SDP

Catalog No. 18620756 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-620-756 100 g
18-620-755 25 g
18-620-757 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-620-756 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. F1272100GM
Add to Cart
Add to Cart

Synonym: Fluorescein Disodium, Acid Yellow 73 Sodium Salt, Resorcinolphthalein Sodium

Fluorescein Sodium is a synthetic organic compound that is a common fluorochrome dye. It is obtained from the condensation of phthalic anhydride with resorcinol on heating. It is used in dying silk and wool, ink manufacturing, in coloring cosmetics, and in coloring gasoline and as a toner. It is widely used as a fluorescent tracer for many applications. It appears as a yellowish to red solid crystalline powder that is soluble in acetone, methanol, hot alcohol, glacial acetic acid, alkali hydroxides, carbonates and water. Ungraded products supplied by Spectrum™ are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Chemical Identifiers

CAS 518-47-8

Specifications

Packaging Amber Glass
Color Orange to Red
Quantity 100 g
Chemical Name or Material Fluorescein Sodium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.