Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluorescein, Water Soluble CI 45350 (Acid Yellow 73; Sodium Fluorescein), Reagents

Catalog No. 01336755
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-336-755 25 g
01-336-756 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-336-755 Supplier Reagents Holdings Llc Supplier No. C611100025C1
Add to Cart
Add to Cart

Fluorescein, Water Soluble CI 45350 (Acid Yellow 73; Sodium Fluorescein), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 518-47-8
Molecular Formula C20H10Na2O5
Molecular Weight (g/mol) 376.28
MDL Number MFCD00167039
InChI Key NJDNXYGOVLYJHP-UHFFFAOYSA-L
Synonym Disodium Fluorescein, Uranine, Color Index 45350
IUPAC Name disodium 2-(6-oxido-3-oxo-3H-xanthen-9-yl)benzoate
SMILES [Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=CC(=O)C=C2OC2=CC([O-])=CC=C12

Specifications

Grade Reagent
Packaging Glass Bottle
Specific Gravity 1
Color Brown or Orange to Red
Quantity 25 g
Physical Form Solid
Chemical Name or Material Fluorescein, Water Soluble CI 45350 (Acid Yellow 73, Sodium Fluorescein)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.