Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluorescein, Water Soluble, Spectrum™ Chemical
SDP

Catalog No. 18610604 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Plastic Bottle
Poly Pail
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-610-604 500 g Amber Glass Bottle
18-610-608 25 g Amber Glass Bottle
18-610-602 100 g Amber Glass Bottle
18-610-605 500 g Plastic Bottle
18-610-607 2.5 kg Poly Pail
18-610-603 12 kg Poly Pail
18-610-606 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18-610-604 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. FL116500GMGL
Add to Cart
Add to Cart

Chemical Identifiers

CAS 518-47-8

Specifications

CAS Max % 1
Grade Reagent
Packaging Amber Glass Bottle
Color Orange
Quantity 500 g
Chemical Name or Material Fluorescein

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.