Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Tris-Glycine Running Buffer for SDS-PAGE

Catalog No. ICN08W00010 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
10 Packets

Powder buffer salts for making SDS-PAGE buffer.

  • Comes in powder form
  • 1 packet makes up 500 mL
TRUSTED_SUSTAINABILITY

Specifications

Concentration 1X
For Use With (Application) Western Blotting
For Use With (Equipment) SDS-Page
Physical Form Powder
Quantity 10 Packets
Solution Type Buffer
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Unless specified otherwise, MP Biomedical™'s products are for research or further manufacturing use only, not for direct human use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.