Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluoride Standard, 0.5 ppm F- (Premixed with TISAB), Ricca Chemical
SDP

Catalog No. 3161532 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
3161532 1 L Natural Poly Bottle
3161516 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 3161532 Supplier Ricca Chemical Company Supplier No. 3161.532
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7681-49-4 , 1310-73-2 , 64-19-7 , 7647-14-5 , 13291-61-7 , 7681-49-4
Molecular Formula FNa
Molecular Weight (g/mol) 41.99
MDL Number MFCD00003524
InChI Key PUZPDOWCWNUUKD-UHFFFAOYSA-M
PubChem CID 5235
ChEBI CHEBI:28741
IUPAC Name sodium fluoride
SMILES [F-].[Na+]

Specifications

Packaging Natural Poly Bottle
CAS Min % 93.0, 1.95, 1.64, 1.59, 0.11
CAS Max % 96.0, 2.03, 1.7, 1.65, 0.11
Physical Form Liquid
Color Colorless
Melting Point Approximately 0°C
Quantity 1 L
Concentration or Composition (by Analyte or Components) 1mL = 0.0005mg F-, 0.5ppm F-, Premixed with TISAB
Chemical Name or Material Fluoride Standard
ShelfLife 12 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.