Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluoride Standard, 10 ppm F- (Premixed with TISAB/Color Coded Orange), Ricca Chemical
SDP

Catalog No. 31651 Shop All Ricca Chemical Company Products
Click to view available options
Quantity:
500 mL
1 L
4 L

Specifications

Chemical Name or Material Fluoride Standard
Packaging Natural Poly Bottle
CAS 7681-49-4, 1310-73-2, 64-19-7, 7647-14-5, 13291-61-7, 7732-18-5
CAS Min % 93.0, 1.85, 1.55, 1.5, 0.1, 7.0E-4
CAS Max % 97.0, 1.93, 1.61, 1.56, 0.11, 7.0E-4
Equivalence 1 mL = 0.01 mg F
Physical Form Liquid
Color Orange
Melting Point Approximately 0°C
Quantity 4 L
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.