Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fluoride Standard, 10 ppm F- (Premixed with TISAB/Color Coded Orange), Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
500 mL
1 L
4 L
Specifications
Specifications
Chemical Name or Material | Fluoride Standard |
Packaging | Natural Poly Bottle |
CAS | 7681-49-4, 1310-73-2, 64-19-7, 7647-14-5, 13291-61-7, 7732-18-5 |
CAS Min % | 93.0, 1.85, 1.55, 1.5, 0.1, 7.0E-4 |
CAS Max % | 97.0, 1.93, 1.61, 1.56, 0.11, 7.0E-4 |
Equivalence | 1 mL = 0.01 mg F |
Physical Form | Liquid |
Color | Orange |
Melting Point | Approximately 0°C |
Quantity | 1 L |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction