Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluorinert™ Heptacosafluorotributylamine, MilliporeSigma™ Supelco™

Catalog No. p-7187840 Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
5 mL
25 mL

Catalog No. 11-101-3898

Supplier: MilliporeSigma Supelco 772995ML

Only null left
Add to Cart
Add to Cart
Cleaning Products, Cosmetics, Environmental, Flavors and Fragrances, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Product Line Fluorinert
CAS 311-89-7
Vapor Pressure 1.3 mmHg (25°C)
Vapor Density 23.3 (vs. air)
Density 1.883 g/mL (at 25°C)
Physical Form Neat
Linear Formula N(CF2 CF2 CF2 CF3)3
Technique Mass Spectrometry
MDL Number MFCD00000436
Boiling Point 178°C
Synonym Fluorinert(TM) FC-43; PFTBA; Perfluorotributylamine; Tris(nonafluorobutyl)amine
Type Analytical Standard
Quantity 5 mL
Formula Weight 671.09
Grade Analytical Standard
Show More Show Less

Fluorinert is a trademark of 3M Company

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.