Learn More
Eisco™ Fluorite Mineral Specimen
SCI_ED
Catalog No. S26645 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1 Ea.
10/Pk.
12/Pk.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S26645 10/Pk.
S24685 1 Ea.
S24686 12/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S26645 Supplier Eisco™ Supplier No. ESNG0014PK10
Add to Cart
Add to Cart
Fluorite Mineral Specimen is used in classroom identification lab exercises and for study of uncommon 4-plane cleavage pattern.
  • For use in lessons on Bowen's reaction series
  • Has Mohs scale hardness of 4
  • For lessons about hardness and cleavage, as well as the fickle method of identifying minerals by color
  • Correlates well to Next Generation Science Standards Evidence Statements
Specifications
Product Type Mineral
Quantity 10/Pk.
Specimen Size 2 to 3 cm
Weight (Metric) 0.2 kg
Specimen Type Fluorite