Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Fluoroquinolone 3 Monoclonal Antibody (3E4)
GREENER_CHOICE

Catalog No. PIMA549625
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA549625 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA549625 Supplier Invitrogen™ Supplier No. MA549625
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Antibody tested in indirect ELISA.

Fluoroquinolones are a class of antibiotics that are commonly used to treat a variety of bacterial infections. They work by inhibiting the activity of enzymes that are essential for bacterial DNA replication, thereby preventing the bacteria from multiplying and causing further infection. Fluoroquinolone 3 is a specific gene target that is involved in the development of resistance to fluoroquinolone antibiotics. By targeting this gene, researchers hope to develop new antibiotics that are more effective against resistant strains of bacteria.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fluoroquinolone 3
Applications ELISA, Immunohistochemistry, Lateral Flow, SDS-Page
Classification Monoclonal
Clone 3E4
Concentration 4.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Host Species Mouse
Immunogen Conjugate of FOS(III) hapten and KLH.
Purification Method Protein G
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.