Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fluorosilicic Acid Standard, 600 ppm, Reagents
Click to view available options
Quantity:
4 L
Specifications
Specifications
| Chemical Name or Material | Fluorosilicic Acid Standard, 600 ppm |
| Packaging | Poly Bottle |
| CAS | 7732-18-5, 1333-82-0, 7664-93-9 |
| Specific Gravity | 1.257 |
| Physical Form | Liquid |
| Color | Colorless |
| Boiling Point | 100°C |
| Solubility | Completely miscible |
| Quantity | 4 L |
| Grade | Reagent |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction